Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161044.1 | NP_001154516.1 | 2 - 3 | 747 | 99 | |
NM_001335415.1 | NP_001323471.1 | 2 - 3 | 776 | 99 | ||
NM_126984.4 | NP_565362.1 | 2 - 3 | 902 | 99 | ||
NM_126985.6 | NP_565363.2 | 2 - 3 | 747 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |