Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100149.4 | NP_171769.1 | 1 - 2 | 488 | 87 | |
NM_202014.1 | NP_973743.1 | 1 - 2 | 362 | 87 | ||
GenBank mRNA | AY070068.1 | - | 1 - 2 | 362 | 87 | |
AY096437.1 | - | 1 - 2 | 225 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |