Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100396.4 | NP_172010.1 | 13 - 14 | 1713 | 68 | |
NM_202032.2 | NP_973761.1 | 14 - 15 | 1486 | 68 | ||
GenBank mRNA | AY050379.1 | - | 13 - 14 | 1573 | 68 | |
L13922.1 | - | 13 - 14 | 1552 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |