Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL5-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331745.1 | NP_001318951.1 | 3 - 4 | 314 | 73 | |
NM_100670.2 | NP_172274.1 | 3 - 4 | 280 | 73 | ||
NM_202058.2 | NP_973787.1 | 2 - 3 | 672 | 73 | ||
NM_202059.3 | NP_973788.1 | 3 - 4 | 291 | 73 | ||
GenBank mRNA | AK117714.1 | - | 3 - 4 | 281 | 73 | |
BT004994.1 | - | 2 - 3 | 180 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |