Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100780.3 | NP_172383.3 | 10 - 11 | 2492 | 107 | |
NM_202070.1 | NP_973799.1 | 11 - 12 | 2418 | 107 | ||
GenBank mRNA | BT005716.1 | - | 10 - 11 | 2493 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |