Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332007.1 | NP_001321930.1 | 7 - 8 | 973 | 69 | |
NM_101058.2 | NP_172651.1 | 9 - 10 | 1308 | 69 | ||
NM_179316.3 | NP_849647.1 | 9 - 10 | 1403 | 69 | ||
NM_202083.1 | NP_973812.1 | 9 - 10 | 1294 | 69 | ||
GenBank mRNA | AY091115.1 | - | 9 - 10 | 1315 | 69 | |
AY122978.1 | - | 9 - 10 | 1265 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |