Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035969.2 | NP_001031046.1 | 5 - 6 | 706 | 80 | |
NM_001332114.1 | NP_001319001.1 | 5 - 6 | 1051 | 80 | ||
NM_001332115.1 | NP_001323290.1 | 5 - 6 | 870 | 80 | ||
NM_001332116.1 | NP_001323291.1 | 5 - 6 | 931 | 80 | ||
NM_001332117.1 | NP_001323289.1 | 5 - 6 | 800 | 80 | ||
NM_101305.4 | NP_563950.1 | 5 - 6 | 844 | 80 | ||
NM_202095.3 | NP_973824.1 | 5 - 6 | 758 | 80 | ||
GenBank mRNA | AK117145.1 | - | 5 - 6 | 758 | 80 | |
BT005935.1 | - | 4 - 5 | 586 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |