Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101652.3 | NP_564040.1 | 2 | 1144 | 84 | |
NM_202124.2 | NP_973853.1 | 1 | 1268 | 84 | ||
NM_202125.3 | NP_973854.1 | 2 | 1172 | 84 | ||
GenBank mRNA | AK317382.1 | - | 2 | 1120 | 84 | |
AY063880.1 | - | 2 | 1082 | 84 | ||
AY096364.1 | - | 2 | 936 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |