Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036016.2 | NP_001031093.1 | 3 - 4 | 617 | 94 | |
NM_001332707.1 | NP_001323071.1 | 3 - 4 | 854 | 94 | ||
NM_001332708.1 | NP_001323072.1 | 3 - 4 | 1053 | 94 | ||
NM_001332709.1 | NP_001323073.1 | 3 - 4 | 1013 | 94 | ||
NM_102390.3 | NP_173950.1 | 2 - 3 | 1120 | 94 | ||
NM_202184.3 | NP_973913.1 | 3 - 4 | 864 | 94 | ||
GenBank mRNA | AJ630483.1 | - | 3 - 4 | 557 | 94 | |
AK117355.1 | - | 2 - 3 | 820 | 94 | ||
AY568655.1 | - | 3 - 4 | 557 | 94 | ||
BT005334.1 | - | 2 - 3 | 710 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |