Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036017.1 | NP_001031094.1 | 2 - 3 | 343 | 69 | |
NM_001332712.1 | NP_001321599.1 | 1 - 2 | 662 | 69 | ||
NM_102407.2 | NP_564246.1 | 2 - 3 | 344 | 69 | ||
NM_179377.2 | NP_849708.1 | 1 - 2 | 688 | 69 | ||
NM_202186.3 | NP_973915.1 | 2 - 3 | 377 | 69 | ||
GenBank mRNA | AK317434.1 | - | 2 - 3 | 315 | 69 | |
AY058066.1 | - | 1 - 2 | 571 | 69 | ||
BT003016.1 | - | 1 - 2 | 255 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |