Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102576.2 | NP_564298.1 | 7 - 8 | 1085 | 72 | |
NM_202197.2 | NP_973926.1 | 7 - 8 | 1088 | 72 | ||
GenBank mRNA | AY059854.1 | - | 7 - 8 | 1082 | 72 | |
BT008763.1 | - | 7 - 8 | 1021 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |