Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179414.1 | NP_849745.1 | 3 - 4 | 305 | 110 | |
NM_202231.1 | NP_973960.1 | 3 - 4 | 305 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |