Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUVR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337514.1 | NP_001327806.1 | 4 - 5 | 1480 | 96 | |
NM_001337515.1 | NP_001327808.1 | 4 - 5 | 1446 | 96 | ||
NM_001337516.1 | NP_001327807.1 | 5 - 6 | 1381 | 96 | ||
NM_001337517.1 | NP_001327809.1 | 5 - 6 | 1365 | 96 | ||
NM_111309.3 | NP_187088.2 | 4 - 5 | 1467 | 96 | ||
NM_202488.2 | NP_974217.1 | 5 - 6 | 1386 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |