Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337544.1 | NP_001319473.1 | 13 - 14 | 1804 | 94 | |
NM_001337545.1 | NP_001319474.1 | 13 - 14 | 1682 | 94 | ||
NM_111359.4 | NP_187138.1 | 12 - 13 | 1773 | 94 | ||
NM_202493.3 | NP_974222.1 | 13 - 14 | 1669 | 94 | ||
GenBank mRNA | AY059920.1 | - | 12 - 13 | 1634 | 94 | |
AY072096.1 | - | 12 - 13 | 1640 | 94 | ||
AY096583.1 | - | 12 - 13 | 1526 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |