Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337544.1 | NP_001319473.1 | 5 - 6 | 729 | 135 | |
NM_001337545.1 | NP_001319474.1 | 5 - 6 | 607 | 135 | ||
NM_111359.4 | NP_187138.1 | 4 - 5 | 698 | 135 | ||
NM_202493.3 | NP_974222.1 | 5 - 6 | 594 | 135 | ||
GenBank mRNA | AY059920.1 | - | 4 - 5 | 559 | 135 | |
AY072096.1 | - | 4 - 5 | 565 | 135 | ||
AY096583.1 | - | 4 - 5 | 451 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |