Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337544.1 | NP_001319473.1 | 7 - 8 | 910 | 72 | |
NM_001337545.1 | NP_001319474.1 | 7 - 8 | 788 | 72 | ||
NM_111359.4 | NP_187138.1 | 6 - 7 | 879 | 72 | ||
NM_202493.3 | NP_974222.1 | 7 - 8 | 775 | 72 | ||
GenBank mRNA | AY059920.1 | - | 6 - 7 | 740 | 72 | |
AY072096.1 | - | 6 - 7 | 746 | 72 | ||
AY096583.1 | - | 6 - 7 | 632 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |