Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337544.1 | NP_001319473.1 | 9 - 10 | 1236 | 127 | |
NM_001337545.1 | NP_001319474.1 | 9 - 10 | 1114 | 127 | ||
NM_111359.4 | NP_187138.1 | 8 - 9 | 1205 | 127 | ||
NM_202493.3 | NP_974222.1 | 9 - 10 | 1101 | 127 | ||
GenBank mRNA | AY059920.1 | - | 8 - 9 | 1066 | 127 | |
AY072096.1 | - | 8 - 9 | 1072 | 127 | ||
AY096583.1 | - | 8 - 9 | 958 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |