Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337563.1 | NP_001327889.1 | 4 - 5 | 1171 | 72 | |
NM_111386.3 | NP_187165.2 | 3 - 4 | 1228 | 72 | ||
GenBank mRNA | AY081340.1 | - | 3 - 4 | 1228 | 72 | |
AY128831.1 | - | 3 - 4 | 687 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |