Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111402.5 | NP_566251.1 | 2 - 3 | 594 | 126 | |
GenBank mRNA | AY048291.1 | - | 2 - 3 | 459 | 126 | |
BT000838.1 | - | 2 - 3 | 373 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |