Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035566.2 | NP_001030643.1 | 2 - 3 | 293 | 73 | |
NM_001202883.1 | NP_001189812.1 | 2 - 3 | 258 | 73 | ||
NM_001202884.1 | NP_001189813.1 | 3 - 4 | 226 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |