Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111415.5 | NP_187193.3 | 14 - 15 | 1525 | 76 | |
NM_202498.2 | NP_974227.1 | 14 - 15 | 1635 | 76 | ||
GenBank mRNA | AK228046.1 | - | 14 - 15 | 1526 | 76 | |
BT006458.1 | - | 14 - 15 | 1423 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |