Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111415.5 | NP_187193.3 | 3 - 4 | 396 | 60 | |
NM_202498.2 | NP_974227.1 | 3 - 4 | 503 | 60 | ||
GenBank mRNA | AK228046.1 | - | 3 - 4 | 397 | 60 | |
BT006458.1 | - | 3 - 4 | 294 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |