Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202886.1 | NP_001189815.1 | 9 - 10 | 1290 | 83 | |
NM_001337585.1 | NP_001319478.1 | 8 - 9 | 1269 | 83 | ||
NM_001337586.1 | NP_001319479.1 | 8 - 9 | 1255 | 83 | ||
GenBank mRNA | BT029213.1 | - | 8 - 9 | 1158 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |