Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111426.4 | NP_187204.1 | 4 - 5 | 720 | 100 | |
GenBank mRNA | AF123394.1 | - | 4 - 5 | 575 | 100 | |
AY062705.1 | - | 4 - 5 | 646 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |