Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111426.4 | NP_187204.1 | 8 - 9 | 1240 | 63 | |
GenBank mRNA | AF123394.1 | - | 8 - 9 | 1095 | 63 | |
AY062705.1 | - | 8 - 9 | 1166 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |