Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337605.1 | NP_001327100.1 | 5 - 6 | 1004 | 69 | |
NM_111448.3 | NP_187225.2 | 6 - 7 | 1124 | 69 | ||
GenBank mRNA | AJ404470.1 | - | 6 - 7 | 1091 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |