Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337610.1 | NP_001325548.1 | 8 - 9 | 972 | 100 | |
NM_111453.1 | NP_566259.1 | 8 - 9 | 919 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |