Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCI2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111463.2 | NP_187240.1 | 2 - 3 | 560 | 106 | |
GenBank mRNA | AF122006.1 | - | 2 - 3 | 245 | 106 | |
AK176253.1 | - | 2 - 3 | 264 | 106 | ||
AK228716.1 | - | 2 - 3 | 268 | 106 | ||
AY084701.1 | - | 2 - 3 | 270 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |