Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111471.3 | NP_566265.1 | 10 - 11 | 975 | 82 | |
GenBank mRNA | AY094418.1 | - | 10 - 11 | 889 | 82 | |
BT001117.1 | - | 10 - 11 | 834 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |