Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111471.3 | NP_566265.1 | 13 - 14 | 1239 | 113 | |
GenBank mRNA | AY094418.1 | - | 13 - 14 | 1153 | 113 | |
BT001117.1 | - | 13 - 14 | 1098 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |