Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111477.4 | NP_187254.1 | 15 - 16 | 1993 | 94 | |
GenBank mRNA | AB000799.1 | - | 15 - 16 | 1881 | 94 | |
AY069917.1 | - | 15 - 16 | 1975 | 94 | ||
BT001024.1 | - | 15 - 16 | 1840 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |