Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111477.4 | NP_187254.1 | 5 - 6 | 748 | 69 | |
GenBank mRNA | AB000799.1 | - | 5 - 6 | 636 | 69 | |
AY069917.1 | - | 5 - 6 | 730 | 69 | ||
BT001024.1 | - | 5 - 6 | 595 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |