Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337640.1 | NP_001326821.1 | 2 - 3 | 1101 | 108 | |
NM_111494.3 | NP_566275.1 | 2 - 3 | 1134 | 108 | ||
GenBank mRNA | AY045891.1 | - | 2 - 3 | 1103 | 108 | |
AY091449.1 | - | 2 - 3 | 763 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |