Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111524.4 | NP_187300.3 | 5 - 6 | 1325 | 69 | |
GenBank mRNA | AF039406.1 | - | 5 - 6 | 1171 | 69 | |
AY035017.1 | - | 5 - 6 | 1185 | 69 | ||
AY059083.1 | - | 5 - 6 | 1042 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |