Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111530.4 | NP_187306.2 | 1 - 2 | 837 | 150 | |
GenBank mRNA | AF409086.2 | - | 1 - 2 | 814 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |