Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111530.4 | NP_187306.2 | 3 - 4 | 1065 | 78 | |
GenBank mRNA | AF409086.2 | - | 3 - 4 | 1042 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |