Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111530.4 | NP_187306.2 | 5 - 6 | 1361 | 99 | |
GenBank mRNA | AF409086.2 | - | 5 - 6 | 1338 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |