Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111545.3 | NP_187321.2 | 1 - 2 | 242 | 132 | |
NM_202513.3 | NP_974242.1 | 1 - 2 | 355 | 132 | ||
GenBank mRNA | AJ584646.1 | - | 1 - 2 | 125 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |