Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111561.5 | NP_187337.2 | 15 - 16 | 2470 | 76 | |
GenBank mRNA | AY091014.1 | - | 15 - 16 | 2233 | 76 | |
BT002365.1 | - | 15 - 16 | 2179 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |