Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111566.4 | NP_187342.1 | 4 - 5 | 488 | 78 | |
GenBank mRNA | AF123254.1 | - | 4 - 5 | 297 | 78 | |
AY062621.1 | - | 4 - 5 | 354 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |