Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111566.4 | NP_187342.1 | 6 - 7 | 641 | 81 | |
GenBank mRNA | AF123254.1 | - | 6 - 7 | 450 | 81 | |
AY062621.1 | - | 6 - 7 | 507 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |