Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337693.1 | NP_001326712.1 | 14 - 15 | 1706 | 125 | |
NM_001337694.1 | NP_001326713.1 | 13 - 14 | 1574 | 125 | ||
NM_111596.6 | NP_187372.5 | 13 - 14 | 1510 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |