Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337693.1 | NP_001326712.1 | 38 - 39 | 4264 | 102 | |
NM_001337694.1 | NP_001326713.1 | 37 - 38 | 4132 | 102 | ||
NM_111596.6 | NP_187372.5 | 37 - 38 | 4068 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |