Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111616.3 | NP_566305.1 | 3 - 4 | 485 | 118 | |
GenBank mRNA | AY042807.1 | - | 3 - 4 | 485 | 118 | |
AY064647.1 | - | 3 - 4 | 237 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |