Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035582.1 | NP_001030659.1 | 2 - 3 | 736 | 80 | |
NM_001125127.2 | NP_001118599.1 | 2 - 3 | 831 | 80 | ||
NM_111644.5 | NP_187422.1 | 2 - 3 | 824 | 80 | ||
NM_202521.2 | NP_974250.1 | 2 - 3 | 739 | 80 | ||
GenBank mRNA | AK316934.1 | - | 2 - 3 | 756 | 80 | |
BT029490.1 | - | 1 - 2 | 524 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |