Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111667.4 | NP_187445.1 | 2 - 3 | 549 | 96 | |
GenBank mRNA | AF412276.1 | - | 2 - 3 | 307 | 96 | |
AK118247.1 | - | 2 - 3 | 474 | 96 | ||
AY085508.1 | - | 2 - 3 | 503 | 96 | ||
BT005578.1 | - | 2 - 3 | 299 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |