Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111678.3 | NP_187456.1 | 7 - 8 | 1280 | 110 | |
GenBank mRNA | BT015083.1 | - | 7 - 8 | 1189 | 110 | |
BT015904.1 | - | 7 - 8 | 1130 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |