Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337758.1 | NP_001319499.1 | 3 - 4 | 667 | 95 | |
NM_111683.2 | NP_187461.1 | 2 - 3 | 752 | 95 | ||
GenBank mRNA | AF448231.1 | - | 3 - 4 | 566 | 95 | |
AY056439.1 | - | 2 - 3 | 551 | 95 | ||
AY057517.1 | - | 2 - 3 | 551 | 95 | ||
AY133652.1 | - | 2 - 3 | 449 | 95 | ||
BT006370.1 | - | 2 - 3 | 449 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |