Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111689.5 | NP_187467.1 | 10 - 11 | 1831 | 97 | |
GenBank mRNA | AF543710.1 | - | 10 - 11 | 1353 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |