Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111690.2 | NP_187468.1 | 4 - 5 | 697 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |